Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588054 Supplier Invitrogen™ Supplier No. PA588054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGVLFHSSPA LQPAADHKPG PGARAEDAAE GRARRREEGA PGDPEAALED NLARIRENHE RALREAKETL QKLPEEIQRD ILLEKKKVAQ DQLRDKAPFR GLPPVDFVPP IGVESREPAD A; Positive Samples: 293T, LO2, Raji, A-549; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the hydrolysis of three terminal mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAN1A1
Gene Accession No. P33908
Gene Alias Aa2-166; Alpha-1,2-mannosidase IA; HUMM3; HUMM9; Man(9)-alpha-mannosidase; Man1a; MAN1A1; MAN9; Man9-mannosidase; mannosidase 1, alpha; mannosidase alpha class 1A member 1; mannosidase, alpha, class 1A, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase 1A; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA-like; mannosyl-oligosaccharide alpha-1,2-mannosidase; PCR1; processing alpha-1,2-mannosidase IA; si:dkey-164k19.1; si:dkeyp-6c7.1
Gene Symbols MAN1A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-190 of human MAN1A1 (NP_0058982).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4121
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.