Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100606 Supplier Invitrogen™ Supplier No. PA5100606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAN1A2.

Alpha-mannosidases function at different stages of N-glycan maturation in mammalian cells. See MAN2A1 (MIM 154582) for general information.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAN1A2
Gene Accession No. O60476
Gene Alias 3.2.1.113; AI428775; AI528764; AI854422; alpha1,2-mannosidase; Alpha-1,2-mannosidase IB; Man1a2; MAN1B; mannosidase 1, beta; mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; PCR2; Processing alpha-1,2-mannosidase IB
Gene Symbols MAN1A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAN1A2(Accession O60476), corresponding to amino acid residues K297-S347.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10905
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.