Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589717 Supplier Invitrogen™ Supplier No. PA589717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSKHKRFDLG LEDVLIPHVD AGKGAKNPGV FLIHGPDEHR HREEEERLRN KIRADHEKAL EEAKEKLRKS REEIRAEIQT EKNKVVQEMK IKENKPLPPV PIPNLVGIRG GDPEDNDIRE K; Positive Samples: LO2, A-549, Mouse brain, Mouse liver; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

Alpha-mannosidases function at different stages of N-glycan maturation in mammalian cells. See MAN2A1 (MIM 154582) for general information.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAN1A2
Gene Accession No. O60476, P39098
Gene Alias 3.2.1.113; AI428775; AI528764; AI854422; alpha1,2-mannosidase; Alpha-1,2-mannosidase IB; Man1a2; MAN1B; mannosidase 1, beta; mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; PCR2; Processing alpha-1,2-mannosidase IB
Gene Symbols MAN1A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human MAN1A2 (NP_0066901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10905, 17156
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.