Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN1A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556974
Description
Immunogen sequence: KLRKSREEIR AEIQTEKNKV VQEMKIKENK PLPPVPIPNL VGIRGGDPED NDIREK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
Alpha-mannosidases function at different stages of N-glycan maturation in mammalian cells. See MAN2A1 (MIM 154582) for general information.
Specifications
MAN1A2 | |
Polyclonal | |
Unconjugated | |
MAN1A2 | |
3.2.1.113; AI428775; AI528764; AI854422; alpha1,2-mannosidase; Alpha-1,2-mannosidase IB; Man1a2; MAN1B; mannosidase 1, beta; mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; PCR2; Processing alpha-1,2-mannosidase IB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10905 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.40 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60476 | |
MAN1A2 | |
Recombinant protein corresponding to Human MAN1A2. Recombinant protein control fragment (Product #RP-95731). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction