Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAN1A2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501729175 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9175 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9175 Supplier Proteintech Group Inc Supplier No. 140491AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the maturation of Asn-linked oligosaccharides. Progressively trim alpha-1, 2-linked mannose residues from Man(9)GlcNAc(2) to produce Man(5)GlcNAc(2).

Specifications

Antigen MAN1A2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAN1A2
Gene Accession No. O60476
Gene Alias Alpha 1,2 mannosidase IB, MAN1A2, MAN1B
Gene Symbols MAN1A2
Host Species Rabbit
Immunogen MAN1A2 Fusion Protein Ag5240
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10905
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.