Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562428 Supplier Invitrogen™ Supplier No. PA562428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: METGLSPEIV HFNLYPQPGR RDVEVKPADR HNLLRPETVE SLFYLYRVTG DRKYQDWGWE ILQSFSRFTR VPSGGYSSIN NVQDPQKP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Involved in glycoprotein quality control targeting of misfolded glycoproteins for degradation. It primarily trims a single alpha-1,2-linked mannose residue from Man9GlcNAc2 to produce Man8GlcNAc2, but at high enzyme concentrations, as found in the ER quality control compartment (ERQC), it further trims the carbohydrates to Man5-6GlcNAc2. Tremblay L.O., Glycobiology 9:1073-1078(1999) [PubMed: 10521544]. Gonzalez D.S., J. Biol. Chem. 274:21375-21386(1999) [PubMed: 10409699]. Clark H.F., Genome Res. 13:2265-2270(2003) [PubMed: 12975309].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAN1B1
Gene Accession No. Q9UKM7
Gene Alias 3.2.1.113; E430019H13Rik; endoplasmic Reticulum Class I alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; ERManI; Gm108; MAN1B1; Man9GlcNAc2-specific processing alpha-mannosidase; man9GlcNAc2-specific-processing alpha-mannosidase; MANA-ER; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1; MRT15; RGD1563595; RP11-350O14.2; UNQ747/PRO1477
Gene Symbols MAN1B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAN1B1. Recombinant protein control fragment (Product #RP-101484).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.