Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1C1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA588794

Catalog No. PIPA588794


Only null left
Add to Cart

Description

Description

Immunogen sequence: PHSSRLKRLF LAPRTQQPGL EVVAEIAGHA PAREQEPPPN PAPAAPAPGE DDPSSWASPR RRKGGLRRTR PTGPREEATA ARGNSIPASR PGDEGVPFRF DFNAFRSRLR HPVLGTRADE SQEPQSQVRA QREKIKEMMQ FAWQSYKRYA MGKNELRPLT KDGYEGNMFG GLSGATVIDS LDTLYLMELK EEFQEAK; Positive Samples: A-549, U-87MG, HepG2, Mouse liver; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

MAN1C1 gene ontology annotations related to this gene include metabolic process.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAN1C1
Polyclonal
Unconjugated
MAN1C1
1,2-alpha-mannosidase IC; AI593348; Alpha-1,2-mannosidase IC; HMIC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; MAN1A3; MAN1C; MAN1C1; Mannosidase alpha class 1C member 1; mannosidase, alpha, class 1C, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; pp6318; processing alpha-1,2-mannosidase IC
Rabbit
Affinity chromatography
RUO
230815, 57134
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.04 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q9NR34
MAN1C1
Recombinant fusion protein containing a sequence corresponding to amino acids 44-240 of human MAN1C1 (NP_0651121).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.