Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN2A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582944 Supplier Invitrogen™ Supplier No. PA582944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVVYNPLEQD RISLVSVYVS SPTVQVFSAS GKPVEVQVSA VWDTANTISE TAYEISFRAH IPPLGLKVYK ILESASSNSH LADYVLYKNK VEDSGIFTIK NMINTEEGIT LENSFVLLRF DQTGLMKQMM TKED Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 63%.

This gene encodes a glycosyl hydrolase that localizes to the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN2A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAN2A1
Gene Accession No. Q16706
Gene Alias Alpha-mannosidase 2; alpha-mannosidase II; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; GOLIM7; man II; Man2a1; Mana2; Mana-2; MANII; Manno; mannosidase 2, alpha 1; mannosidase alpha 2; mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; Map-2
Gene Symbols MAN2A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAN2A1. Recombinant protein control fragment (Product #RP-90113).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4124
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.