Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN2A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585045 Supplier Invitrogen™ Supplier No. PA585045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLPSSVRIYL HGRQLSVSRH EAFPLRVIDS GTSDFALSNR YMQVWFSGLT GLLKSIRRVD EEHEQQVDMQ VLVYGTRTSK D Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 79%.

Catalyzes the first committed step in the biosynthesis of complex N-glycans. It controls conversion of high mannose to complex N-glycans; the final hydrolytic step in the N-glycan maturation pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN2A2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAN2A2
Gene Accession No. P49641
Gene Alias 1700052O22Rik; 4931438M07Rik; AI480988; alpha mannosidase IIx; alpha-mannosidase 2x; Alpha-mannosidase IIx; MAN IIx; Man2a2; Mana2x; mannosidase 2, alpha 2; mannosidase alpha class 2A member 2; mannosidase, alpha type II-X; mannosidase, alpha, class 2A, member 2; mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; manosidase, alpha-, type II, isozyme X; MX
Gene Symbols MAN2A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAN2A2. Recombinant protein control fragment (Product #RP-108694).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.