Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN2A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120673
This item is not returnable.
View return policy
Description
Immunogen sequence: VPPEPRPSFF SISPQDCQFA LGGRGQKPEL QMLTVSEELP FDNVDGGVWR QGFDISYDPH DWDAEDLQVF VVPHSHNDPG WIKTFDKYYT EQTQHILNSM V.
Catalyzes the first committed step in the biosynthesis of complex N-glycans. It controls conversion of high mannose to complex N-glycans; the final hydrolytic step in the N-glycan maturation pathway.
Specifications
| MAN2A2 | |
| Polyclonal | |
| Unconjugated | |
| MAN2A2 | |
| 1700052O22Rik; 4931438M07Rik; AI480988; alpha mannosidase IIx; alpha-mannosidase 2x; Alpha-mannosidase IIx; MAN IIx; Man2a2; Mana2x; mannosidase 2, alpha 2; mannosidase alpha class 2A member 2; mannosidase, alpha type II-X; mannosidase, alpha, class 2A, member 2; mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; manosidase, alpha-, type II, isozyme X; MX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4122 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P49641 | |
| MAN2A2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MAN2A2 (NP_006113.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction