Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAN2A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | MAN2A2 |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MAN2A2 Polyclonal antibody specifically detects MAN2A2 in Human samples. It is validated for Western BlotSpecifications
MAN2A2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cellular Markers, Golgi Apparatus Markers | |
PBS, 50% glycerol, pH7.3 | |
4122 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
alpha-mannosidase 2x, alpha-mannosidase IIx, MAN IIx, MANA2X, mannosidase, alpha type II-X, mannosidase, alpha, class 2A, member 2, mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase, manosidase, alpha-, type II, isozyme X | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MAN2A2 (NP_006113.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title