Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAN2B2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297471MFV610
Description
MAN2B2 Polyclonal antibody specifically detects MAN2B2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MAN2B2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
mannosidase, alpha, class 2B, member 2 | |
Produced in rabbits immunized with E. coli-derived Human MAN2B2 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
23324 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction