Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN2B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557911
Description
Immunogen sequence: LLCHRIEQEY QAGPLELNRE AVLRTSTNLN SQQVIYSDNN GYQMQRRPYV SYVNNSIARN YYPMVQSAFM EDGKSRLVLL SERAHGISSQ GNGQVE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Specifications
MAN2B2 | |
Polyclonal | |
Unconjugated | |
MAN2B2 | |
135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Ab2-450; core-specific lysosomal alpha-1,6-Mannosidase; epididymis-specific alpha-mannosidase; HMG17L3; Kiaa0935; Man2b2; mannosidase 2, alpha B2; mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; mKIAA0935; NHC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2E5 | |
MAN2B2 | |
Recombinant protein corresponding to Human MAN2B2. Recombinant protein control fragment (Product #RP-96590). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction