Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN2B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113537
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113537 Supplier Invitrogen™ Supplier No. PA5113537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAN2B2 Polyclonal Antibody for ELISA, IHC (P), Western Blot

The MAN2B2 (Mannosidase Alpha Class 2B Member 2) gene encodes an enzyme that is involved in the processing and modification of glycoproteins by hydrolyzing alpha-mannosidic linkages. Located on chromosome 4, the MAN2B2 protein is crucial for proper lysosomal function and is part of the glycosyl hydrolase family. Pathogenic variants in the MAN2B2 gene have been linked to a rare congenital disorder of glycosylation (CDG), known as MAN2B2-CDG, characterized by immune deficiency, developmental delay, stroke, and dysmorphic facial features. These mutations lead to an altered mannosylation profile, impacting glycoprotein processing and cellular functions. Studies have shown that reintroducing wild-type MAN2B2 can restore the normal mannosylation profile in patient cells, highlighting its therapeutic potential. Furthermore, recent research has identified circular RNA from this gene (circMAN2B2) that is implicated in the proliferation of hepatocellular carcinoma cells, suggesting a role in cancer biology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN2B2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MAN2B2
Gene Accession No. Q9Y2E5
Gene Alias 135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Ab2-450; core-specific lysosomal alpha-1,6-Mannosidase; epididymis-specific alpha-mannosidase; HMG17L3; Kiaa0935; Man2b2; mannosidase 2, alpha B2; mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2; mKIAA0935; NHC
Gene Symbols MAN2B2
Host Species Rabbit
Immunogen Recombinant Human Epididymis-specific alpha-mannosidase protein (347-523AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23324
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.