Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN2C1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566917
Description
Immunogen sequence: LGKDNQRSFQA LYTANQMVNV CDPAQPETFP VAQALASRFF GQHGGESQHT IHATGHCHID TAWLWPFKET VRKCARSWVT ALQLMERNP Highest antigen sequence identity to the following orthologs - mouse 84%, rat 88%.
Hydrolysis of terminal, non-reducing alpha-D-mannose residues in alpha-D-mannosides. Target protein plays a role in the processing of asparagine-linked oligosaccharides.
Specifications
MAN2C1 | |
Polyclonal | |
Unconjugated | |
MAN2C1 | |
alpha mannosidase 6A8B; Alpha-D-mannoside mannohydrolase; alpha-mannosidase 2C1; MAN2C1; MAN6A8; MANA; MANA1; Mannosidase alpha class 2C member 1; mannosidase, alpha 6A8; mannosidase, alpha A, cytoplasmic; mannosidase, alpha, class 2C, member 1; testicular tissue protein Li 115 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NTJ4 | |
MAN2C1 | |
Recombinant Human MAN2C1. Recombinant protein control fragment (Product #RP-107570). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction