Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAN2C1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549241
Description
Hydrolysis of terminal, non-reducing alpha-D-mannose residues in alpha-D-mannosides.
Specifications
MAN2C1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9NTJ4 | |
MAN2C1 | |
KLH conjugated synthetic peptide between 113-142 amino acids from the N-terminal region of human MAN2C1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAN2C1 | |
Alpha-mannosidase 2C1, Alpha mannosidase 6A8B, Alpha-D-mannoside mannohydrolase, Mannosidase alpha class 2C member 1, MAN2C1, MANA, MANA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4123 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction