Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MANBA Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299160MFV450
Description
MANBA Polyclonal antibody specifically detects MANBA in Human samples. It is validated for Western BlotSpecifications
MANBA | |
Polyclonal | |
Western Blot | |
beta-mannosidase, EC 3.2.1.25, Lysosomal beta A mannosidase, MANB1mannanase, Mannanase, Mannase, mannosidase, beta A, lysosomal | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MANBA | |
0.1 mL | |
Cancer, Endocrinology, Neuroscience, Signal Transduction | |
4126 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction