Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MANBA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590351 Supplier Invitrogen™ Supplier No. PA590351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAELSYSLRG NWSICNGNGS LELPGAVPGC VHSALFQQGL IQDSYYRFND LNYRWVSLDN WTYSKEFKIP FEISKWQKVN LILEGVDTVS KILFNEVTIG ETDNMFNRYS FDITNVVRDV NSIELRFQSA VLYAAQQSKA HTRYQVPPDC PPLVQKGECH VNFVRKEQCS FSWDWGPSFP TQGIWKDVRI EAYNICHLNY FTFSPIYDKS AQEWNLEIES TFDVVSSKPV GGQVIVAIPK LQTQQTYSIE LQPGKRIVEL FVNISKNITV ETWWPHGHGN QTG; Positive Samples: Mouse lung, Mouse testis; Cellular Location: Lysosome.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MANBA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MANBA
Gene Accession No. O00462, Q8K2I4
Gene Alias 2410030O07Rik; B930014J03Rik; beta-mannosidase; beta-mannosidase A; Bmn; Lysosomal beta A mannosidase; MANB1; Manba; mannanase; Mannase; mannosidase beta; mannosidase, beta A, lysosomal
Gene Symbols MANBA
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110173, 4126
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.