Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MANBA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30480320UL
Description
MANBA Polyclonal specifically detects MANBA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MANBA | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
4126 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
beta-mannosidase, EC 3.2.1.25, Lysosomal beta A mannosidase, MANB1mannanase, Mannanase, Mannase, mannosidase, beta A, lysosomal | |
Recombinant fusion protein containing a sequence corresponding to amino acids 18-300 of human MANBA (NP_005899.3). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction