Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MANEA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588029 Supplier Invitrogen™ Supplier No. PA588029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPNTATFGAP FGLDLLPELH QRTIHLGKNF DFQKSDRINS ETNTKNLKSV EITMKPSKAS ELNLDELPPL N; Positive Samples: 293T, U-87MG; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

N-glycosylation of proteins is initiated in the endoplasmic reticulum (ER) by the transfer of the preassembled oligosaccharide glucose-3-mannose-9-N-acetylglucosamine-2 from dolichyl pyrophosphate to acceptor sites on the target protein by an oligosaccharyltransferase complex. This core oligosaccharide is sequentially processed by several ER glycosidases and by an endomannosidase (E.C. 3.2.1.130), such as MANEA, in the Golgi. MANEA catalyzes the release of mono-, di-, and triglucosylmannose oligosaccharides by cleaving the alpha-1,2-mannosidic bond that links them to high-mannose glycans (Hamilton et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MANEA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MANEA
Gene Accession No. Q5SRI9
Gene Alias 4932703L02Rik; alpha 1,2-endomannosidase; ENDO; Endo-alpha mannosidase; endomannosidase; glycoprotein endo-alpha-1,2-mannosidase; hEndo; Mandaselin; Manea; mannosidase endo-alpha; mannosidase, endo-alpha; mEndo
Gene Symbols MANEA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-100 of human MANEA (NP_0789172).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79694
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.