Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MANF Antibody (012), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290750APC
Description
MANF Monoclonal antibody specifically detects MANF in Mouse samples. It is validated for ELISASpecifications
MANF | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
012 | |
ELISA | |
Arginine-Rich Protein, arginine-rich, mutated in early stage tumors, ARMET, ARP, mesencephalic astrocyte-derived neurotrophic factor, Protein ARMET | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MANF (Accession#: NP_083379.2; Met1-Leu179). | |
0.1 mL | |
Biologically Active Proteins, Cancer, DNA Repair | |
7873 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction