Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese, 0.1 ppm In Dilute Nitric Acid, Reagents

Catalog No. 01334808
Change view
Click to view available options
Quantity:
4 L
Catalog No. Quantity
01-334-808 4 L
1 options

Catalog No. 01-334-808

Supplier: Reagents Holdings Llc CM0032004A

Only null left
Add to Cart
Add to Cart

Manganese, 0.1 ppm In Dilute Nitric Acid, Reagent, use for Manganese analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Specific Gravity 1.02
Physical Form Liquid
Color Colorless
pH <2
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Manganese Analysis
Quantity 4 L
Grade Reagent
Chemical Name or Material Manganese, 0.1 ppm In Dilute Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.