Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese AA Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural LDPE Bottle |
Concentration or Composition Notes | 1,000ppm Mn |
CAS Min % | 94.0, 3.88, 0.09 |
CAS Max % | 98.0, 4.04, 0.1 |
Equivalence | 1 mL = 1 mg Mn |
Physical Form | Liquid |
Color | Colorless |
pH | < 2 |
Melting Point | Approximately 0°C |
Quantity | 100 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction