Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese acetate tetrahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number ac212560500
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
6156-78-1
C4H14MnO8
245.09
MFCD00062552
CESXSDZNZGSWSP-UHFFFAOYSA-L
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
93021
manganese(2+);diacetate;tetrahydrate
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
6156-78-1
C4H14MnO8
245.09
MFCD00062552
CESXSDZNZGSWSP-UHFFFAOYSA-L
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
93021
manganese(2+);diacetate;tetrahydrate
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color Pink
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Assay Percent Range 21.8 to 23% (Complexometry)
Linear Formula (CH3CO2)2Mn·4H2O
Flash Point >130°C
Merck Index 14,5724
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 245.09
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Manganese acetate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit