Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Carbonate, Powder, 43.5%, Spectrum™ Chemical
SDP

Catalog No. 18611480 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-480 2.5 kg Poly Bottle
18-611-478 125 g Poly Bottle
18-611-483 500 g Poly Bottle
18-611-481 250 lb. Fiber Drum
18-611-479 12 kg Poly Pail
18-611-482 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-480 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11002.5KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 598-62-9

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 43.5%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.