Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn

Catalog No. p-7050560
Click to view available options
Quantity:
250 g
5 kg

Catalog No. AA3860730

Supplier: Thermo Scientific Chemicals 03860730

Only null left
Add to Cart

CAS: 13434-24-7 | C16H30MnO4 | 341.35 g/mol

Used as paint driers. Accelerating curing agent for unsaturated polyester. Also used as a intermediate for dyestuff, pharmaceutical, syntheses material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13434-24-7
Molecular Formula C16H30MnO4
Molecular Weight (g/mol) 341.35
MDL Number MFCD00058698
InChI Key FHRAKXJVEOBCBQ-UHFFFAOYSA-L
Synonym manganese bis 2-ethylhexanoate, manganese 2-ethylhexanoate, manganese 2+ bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, manganese 2+ salt, hexanoic acid, 2-ethyl-, manganese 2+ salt 2:1, hexanoic acid, 2-ethyl-, manganese salt, hexanoic acid, 2-ethyl-, manganese salt 1:?, manganese tallate 2-ethylhexanoate, manganous 2-ethylhexanoate, mmanganese 2-ethylhexanoate
PubChem CID 114521
IUPAC Name 2-ethylhexanoate;manganese(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Mn+2]

Specifications

Color Brown
Density 0.897 g/mL
Flash Point 40°C (104°F)
Odor Petroleum-like
Quantity 250 g
UN Number UN1268
Solubility Information Soluble in organic solvents. Not miscible or difficult to mix with water.
Formula Weight 341.35
Concentration or Composition (by Analyte or Components) 40% w/w in min.eral spirits, 6% Mn
Physical Form Liquid
Chemical Name or Material Manganese(II) 2-ethylhexanoate
Show More Show Less
Hazard Category H226-H304-H340-H350-H372
Hazard Statement GHS H Statement
H304-H361-H226
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P301+P310-P303+P361+P353-P308+P313-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.