Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(II) acetate tetrahydrate, 99+%, for analysis, ACROS Organics™
Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
Specifications
Assay Percent Range | 99% min. (Complexometry) |
Linear Formula | (CH3CO2)2Mn·4H2O |
Merck Index | 14, 5724 |
Solubility Information | Solubility in water: very soluble. Other solubilities: soluble in alcohol |
Formula Weight | 245.09 |
Physical Form | Crystals |
Percent Purity | 99+% |
Grade | Analysis |
Chloride | 20ppm max |
Chlorine (Cl) | 20ppm max. |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
RTECSNumber : AI5775000