Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) acetate tetrahydrate, 99+%, for analysis

Catalog No. AC205665000
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point >300.0°C
Color Pink
pH 6 to 7 (5% soln.)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula (CH3CO2)2Mn·4H2O
Flash Point >130°C
Merck Index 14, 5724
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
IUPAC Name manganese(2+) diacetate tetrahydrate
Formula Weight 245.09
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 1.58
Chemical Name or Material Manganese(II) acetate tetrahydrate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
RTECSNumber AI5775000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.