Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048533
Click to view available options
Quantity:
5 g
25 g

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
IUPAC Name manganese(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color Pink
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula Mn(OOCCH3)2·4H2O
Merck Index 14,5724
Solubility Information Soluble in water,methanol,ethanol,and acetic acid.
Formula Weight 245.09 (173.03 Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Odor Pungent
Density 1.589 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) acetate tetrahydrate
Show More Show Less
Hazard Category H315-H319-H335-H336-H373
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
EINECSNumber 211-334-3
RTECSNumber AI5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.