Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) bromide, 99%, anhydrous

Catalog No. AC207861000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 13446-03-2 | Br2Mn | 214.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
Molecular Formula Br2Mn
Molecular Weight (g/mol) 214.76
MDL Number MFCD00011113

Specifications

Color Pink
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula MnBr2
Merck Index 15, 5790
Solubility Information Solubility in water: soluble
Formula Weight 214.76
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.38
Density 4.3800g/mL
Chemical Name or Material Manganese(II) bromide, Anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
EINECSNumber 236-591-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.