Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) acetate tetrahydrate, 98+%

Catalog No. AC293062500
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle

CAS: 6147-53-1 | C4H6CoO4·4H2O | 249.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H6CoO4·4H2O
Molecular Weight (g/mol) 249.08
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point 140.0°C
Color Pink to Pink-Red
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula (CH3CO2)2Co·4H2O
Fieser 04,99; 16,95
Merck Index 15, 2417
Solubility Information Solubility in water: 380g/L (20°C). Other solubilities: soluble in acids, soluble in alcohols, soluble in pentyl acetate
Formula Weight 249.08
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) acetate tetrahydrate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Suspected of causing genetic defects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May damage fertility.<br/
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Obtain speci
RTECSNumber AG3325000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.