Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) chloride tetrahydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
13446-34-9
Cl2Mn·4H2O
197.91
13446-34-9
Cl2Mn·4H2O
197.91
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnCl2·4H2O
Fieser 07,222; 09,288
Merck Index 15,5793
Solubility Information Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol,insoluble in ether
Formula Weight 197.91
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 50ppm max.
Iron (Fe) 5ppm max.
Lead (Pb) 5ppm max.
Magnesium (Mg) 50ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Substances Reducing Permanganate 0.0005% (as O) max.
Sulfate 50ppm max.
Zinc (Zn) 10ppm max.
Color Pink
Melting Point 58.0°C
Quantity 500g
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger