Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) chloride tetrahydrate, ACS reagent

Catalog No. p-4440145
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC193451000

Supplier: Thermo Scientific Chemicals 193451000

Only null left
Add to Cart

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58.0°C
Quantity 100 g
Linear Formula MnCl2·4H2O
Fieser 07,222; 09,288
Merck Index 15,5793
Solubility Information Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 197.91
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Manganese(II) chloride tetrahydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.