Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) chloride tetrahydrofuran complex (1:2), 97%, Thermo Scientific™

Catalog No. AC390460010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70317-52-1
Molecular Formula C8H16Cl2MnO2
Molecular Weight (g/mol) 270.06
MDL Number MFCD12912454
InChI Key SJFIRQVWOJTTCR-UHFFFAOYSA-L
PubChem CID 72668716
IUPAC Name dichloromanganese;oxolane
SMILES C1CCOC1.C1CCOC1.Cl[Mn]Cl

Specifications

Color Pink
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 97%
Formula Weight 270.06
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Manganese(II) chloride tetrahydrofuran complex (1:2)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.