Alfa Aesar™ Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4716603

Quantity 1g

 View more versions of this product

Catalog No. AA4716603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-33-2
    Formula Weight 308.75
    MDL Number MFCD00016225
    Merck Index 14,5732
    Chemical Name or Material Manganese(II) iodide, Ultra dry

    Specifications

    Formula Weight 308.75
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White to pink to red
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    TSCA TSCA
    Physical Form Beads
    MDL Number MFCD00016225
    Merck Index 14,5732
    Health Hazard 1 Warning
    Density 5.0g/mL
    Mesh Size −10 Mesh
    Odor Odorless
    Sensitivity Light, Air & Moisture Sensitive
    CAS 7790-33-2
    Quantity 1g
    Storage Note 1
    • Ampouled under argon
    Melting Point 638°C
    Chemical Name or Material Manganese(II) iodide, Ultra dry
    Assay 99.99%, (metals basis)

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit