Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) methoxide

Catalog No. p-7049773
Click to view available options
Quantity:
1 g
5 g

CAS: 7245-20-7 | C2H6MnO2 | 117.006 g/mol

Manganese(II) methoxide is used as a biochemical for proteomics research and it is also used in convenient for carboxylating active methylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7245-20-7
Molecular Formula C2H6MnO2
Molecular Weight (g/mol) 117.006
MDL Number MFCD00061476
InChI Key UQIRCVPINUNHQY-UHFFFAOYSA-N
Synonym manganese ii methoxide, acmc-20akkm, dimethoxymanganese ii, manganese ii bis methoxide, manganese 2+ bis methoxide, manganese 2+ ion bis methoxide
PubChem CID 10219413
IUPAC Name manganese(2+);methanolate
SMILES C[O-].C[O-].[Mn+2]

Specifications

Linear Formula Mn(OCH3)2
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol
Formula Weight 117.01
Physical Form Powder
Chemical Name or Material Manganese(II) methoxide
Hazard Category H228-H302-H315-H319-H335
Hazard Statement GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.