Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) nitrate tetrahydrate, 98%

Catalog No. aaa1852130
Click to view available options
Quantity:
250 g
1000 g

CAS: 20694-39-7 | H8MnN2O10 | 251.006 g/mol

Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

Solubility
Soluble in ethanol and water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20694-39-7
Molecular Formula H8MnN2O10
Molecular Weight (g/mol) 251.006
MDL Number MFCD00149789
InChI Key ALIMWUQMDCBYFM-UHFFFAOYSA-N
Synonym manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
PubChem CID 182616
IUPAC Name manganese(2+);dinitrate;tetrahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]

Specifications

Melting Point ∼26°C
Quantity 250 g
Linear Formula Mn(NO3)2·4H2O
UN Number UN2724
Merck Index 14,5733
Solubility Information Soluble in ethanol and water.
Formula Weight 251.01 (178.95 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Manganese(II) nitrate tetrahydrate
Show More Show Less
Hazard Category H272-H302-H314-H373
Hazard Statement GHS H Statement
H272-H314-H318-H373-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: MANGANESE NITRATE
EINECSNumber 233-828-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.