Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) oxalate dihydrate, Mn 30% min

Catalog No. p-7049555
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg

As a paint and varnish drier | CAS: 6556-16-7 | C2H4MnO6 | 178.986 g/mol

Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-16-7
Molecular Formula C2H4MnO6
Molecular Weight (g/mol) 178.986
MDL Number MFCD00150452
InChI Key HDJUVFZHZGPHCQ-UHFFFAOYSA-L
Synonym manganese ii oxalate 2-hydrate
PubChem CID 131882857
IUPAC Name manganese(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Mn+2]

Specifications

Melting Point 100°C (decomposition)
Color White
Density 2.453 g/mL
Odor Odorless
Quantity 5 g
Merck Index 14,5734
Solubility Information Soluble in dilute acids; Slightly soluble in water.
Formula Weight 178.99 (142.96 Anhydrous)
Concentration or Composition (by Analyte or Components) Mn 30% min.
Physical Form Chunks and/or Powder
Chemical Name or Material Manganese(II) oxalate dihydrate
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 211-367-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.