Alfa Aesar™ Manganese(II) oxide, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4444109

Quantity 10g

 View more versions of this product

Catalog No. AA4444109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-43-0
    Molecular Formula MnO
    Formula Weight 70.94
    MDL Number MFCD00016226
    Chemical Name or Material Manganese (II) oxide

    Specifications

    Formula Weight 70.94
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Green
    RTECS Number OP0900000
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00016226
    CAS 1344-43-0
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H312-H315-H319-H335
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder/Chunks
    Molecular Formula MnO
    Density 5.0g/mL
    Health Hazard 1 Warning
    Chemical Name or Material Manganese (II) oxide
    Assay 99.99%, (metals basis)

    GHS H Statement
    H312-H315-H319-H335
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit