Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)

Catalog No. p-7059227
Click to view available options
Quantity:
10 g
50 g

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol

Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15364-94-0
Molecular Formula Cl2H12MnO14
Molecular Weight (g/mol) 361.92
MDL Number MFCD00150255
InChI Key AIFGVOMWPFMOCN-UHFFFAOYSA-L
Synonym manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade
PubChem CID 203855
IUPAC Name manganese(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 165°C (decomposition)
Physical Form Crystalline
Quantity 10 g
UN Number UN1481
Solubility Information Soluble in water,benzene,ethyl acetate
Formula Weight 361.93 (253.84 Anhydrous)
Percent Purity 99.995%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese (II) perchlorate hexahydrate
Show More Show Less
Hazard Category H272
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-390-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.