Learn More
Manganese(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Description
It used in rocket fuel. This is one of the most proven materials for etching of liquid crystal displays and critical electronics applications as well as ore extraction. It is also used for electropolishing/etching of aluminum, molybdenum, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165°C (decomposition) |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1481 |
Solubility Information | Soluble in benzene, ethyl acetate |
Formula Weight | 361.93 (253.84 Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Manganese(II) perchlorate hexahydrate |
Safety and Handling
Hazard Category | H272 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P280-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S. |
EINECSNumber | 237-390-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.