Learn More
Manganese(II) sulfate tetrahydrate, 99% (metals basis)
Description
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.
Solubility
Soluble in water.
Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | MnSO4·4H2O |
| UN Number | UN3077 |
| Solubility Information | Soluble in water. |
| Formula Weight | 223.06 (151.00 Anhydrous) |
| Percent Purity | 99% |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Chemical Name or Material | Manganese(II) sulfate tetrahydrate |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement P260-P314-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Get medical advice/attention if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 232-089-9 |
| RTECSNumber | OP1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.