Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) sulfide, 99.9% (metals basis)

Catalog No. AA1283514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1283514 25 g
AA1283506 5 g
AA1283522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1283514 Supplier Thermo Scientific Chemicals Supplier No. 01283514
Only null left
Add to Cart

CAS: 18820-29-6 | MnS | 87.00 g/mol

it is used as an intermediate in chemical research& pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18820-29-6
Molecular Formula MnS
Molecular Weight (g/mol) 87.00
MDL Number MFCD00014210
InChI Key VCTOKJRTAUILIH-UHFFFAOYSA-N
Synonym manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size
PubChem CID 87809
SMILES [S--].[Mn++]

Specifications

Melting Point 1610°C (decomposition)
Color Green
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information It is soluble in water. 0.47mg/100 mL (18°C).
IUPAC Name manganese(2+) sulfanediide
Formula Weight 87
Appearance Green powder
Density 3.99
Chemical Name or Material Manganese(II) sulfide
Show More Show Less
Hazard Category H301: Toxic if swallowed.
EINECSNumber 242-599-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.