Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(III) oxide, 98%

Catalog No. aa8779136
Click to view available options
Quantity:
500 g
2 kg

CAS: 1317-34-6 | Mn2O3 | 157.873 g/mol

Manganese(III) oxide is used to produce ceramic magnets and semiconductors. Further, it serves as a precursor to lithium manganese oxide. In addition to this, it acts as a useful battery cathode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-34-6
Molecular Formula Mn2O3
Molecular Weight (g/mol) 157.873
MDL Number MFCD00016217
InChI Key GEYXPJBPASPPLI-UHFFFAOYSA-N
Synonym manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
PubChem CID 14824
IUPAC Name oxo(oxomanganiooxy)manganese
SMILES O=[Mn]O[Mn]=O

Specifications

Melting Point 1080°C (decomposition)
Physical Form Powder
Quantity 500 g
Assay Percent Range 98%
Merck Index 14,5737
Solubility Information Soluble in acid and ammonium chloride. Insoluble in water,alcohol and acetone.
Formula Weight 157.86
Percent Purity 98%
Density 4.50 g/mL
Chemical Name or Material Manganese(III) oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a
EINECSNumber 215-264-4
RTECSNumber OP0915000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.