Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(IV) oxide, 80-85%, powder
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 535.0°C |
Color | Gray |
Physical Form | Powder |
Quantity | 2.5 kg |
Assay Percent Range | 80-85% |
Linear Formula | MnO2 |
Fieser | 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 |
Merck Index | 15, 5795 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid |
Formula Weight | 86.94 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if inhaled. Harmful if swallowed. May cause damage to organs through prolonged or repeated exposure if inhaled. |
Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. D |
EINECSNumber | 215-202-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction