Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(IV) oxide, activated, tech., Mn 58% min

Catalog No. AA1434036
Click to view available options
Quantity:
10 g
100 g
500 g

Catalog No. AA1434036

Supplier: Thermo Scientific Chemicals 01434036

Only null left
Add to Cart

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagentManganese(IV) oxide is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Color Black
Physical Form Powder
Quantity 500 g
Merck Index 14,5730
Solubility Information Insoluble in water,nitric acid,sulfuric acid; Soluble in HCl; In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4
Formula Weight 86.94
Grade Technical
Concentration or Composition (by Analyte or Components) Mn 58% min.
Density 5.026 g/mL
Chemical Name or Material Manganese(IV) oxide, Activated
Show More Show Less
Hazard Category H302+H332-H373
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
EINECSNumber 215-202-6
RTECSNumber OP0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.