Learn More
Manganese(IV) oxide, activated, tech., Mn 58% min
Description
As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagentManganese(IV) oxide is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
Specifications
Specifications
| Melting Point | 535°C (decomposition) |
| Color | Black |
| Physical Form | Powder |
| Quantity | 100 g |
| Merck Index | 14,5730 |
| Solubility Information | Insoluble in water,nitric acid,sulfuric acid; Soluble in HCl; In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4 |
| Formula Weight | 86.94 |
| Grade | Technical |
| Concentration or Composition (by Analyte or Components) | Mn 58% min. |
| Density | 5.026 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H373 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c |
| EINECSNumber | 215-202-6 |
| RTECSNumber | OP0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.