Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Manganese phosphide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1402009

Quantity 10g

 View more versions of this product

Catalog No. AA1402009


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12263-33-1
226.75
MFCD00064736
Manganese phosphide

Specifications

12263-33-1
226.75
−100 Mesh
1095°C
Manganese phosphide
Powder
MFCD00064736
Odorless
10g
99%, (metals basis)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit