Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese phosphide, 99% (metals basis)

Catalog No. p-7049329
Click to view available options
Quantity:
2 g
10 g

CAS: 12263-33-1 | Mn3P2 | 226.76 g/mol

Manganese phosphide is used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12263-33-1
Molecular Formula Mn3P2
Molecular Weight (g/mol) 226.76
MDL Number MFCD00064736
InChI Key ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
IUPAC Name trimanganese(2+) diphosphanetriide
SMILES [P-3].[P-3].[Mn++].[Mn++].[Mn++]

Specifications

Melting Point 1095°C
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 226.75
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Manganese phosphide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.