Learn More
Cytiva Manifold Kit for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
GSA_VA
Catalog No. 45002613 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Catalog No. For Use With (Equipment)
45-002-613 Ettan IPGphor 3 IEF Unit
1 options
Catalog No. 45-002-613 Supplier Cytiva Supplier No. 80649838
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote
Includes
Manifold Kits include: 1 Cleaning brush, 2 Electrode assemblies, 1 Spirit level, 20 Sample cups, 1 Sample cup insertion tool, 60 Paper electrode wicks, 60 Paper bridge pads, 1 Manifold (ceramic or plastic), 1 Forcep, 1 60mL cleaning solution and, User Manual
Manifold Kit for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
Manifold Kit
  • Simplify isoelectric focusing of multiple samples
  • Speed up processing of micropreparative loads in first dimension analysis
  • Handle from 1 to 12 IPG strips simultaneously
  • Sample equilibration can be performed in same tray
  • Efficient for paper-bridge loading
  • Gel capacity: maximum of 12 strips, number of gels: 1 to 12
  • Manifold light available in easy to handle polymeric material for routine general screening applications
  • Ceramic Manifold exhibits uniform thermal characteristics designed for stringent applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For the most stringent applications
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type Ettan IPGphor Manifold, Complete
Includes 1 x Cleaning brush, 2 x Electrode assembly, 1 x Spirit level, 20 x Sample cup, 1 x Sample cup insertion tool, 60 x Paper electrode wicks, 60 x Paper bridge pads, 1 x Manifold , 1 x Forceps, 1 x 60 mL cleaning solution and, User Manual
Material Ceramic
No. of Gels 1 to 12
Quantity 1 Piece